Learn More
Thermo Scientific™ Microscope Slides, Cut, Square
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand:  Thermo Scientific™ AAAC0590059002EB0MNZ#0#G#
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 13487486
  • £435.12 / Pack of 1000
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
59mm
1mm
Uncoated
100 pieces in paper
59 x 59mm
59mm
Extra White
Soda-lime glass
No
Videos