DuPont™ Microporous film Hooded Coverall
Overview
Brand: DuPont™
Manufacturer Part Number: D13483649
X100 COVERALL PROSHIELD 30 SZ S
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11753123
Quantity Price
1 £ 585.0 / Pack of 100
Estimated Shipment
07-03-2017
Add to basket
Description and Specification
Specification
Size Small
Size (Metric) Chest 84 to 92cm
Protection Class Category III, Type 5 and 6, EN 1073-2
Waist Style Elastic
Style Hooded
Product Type Hooded Coverall
Material Microporous Film
Color White
Ankle Style Elastic
For Use With (Application) For that are less demanding in terms of barrier, durability and comfort, for example general maintenance, hospitals and other industries. Spraying or any work involving composite materials, glass manufacturing, and utilities
Seam Type Stitched (internal)