DuPont™ Microporous film Hooded Coverall
Overview
Brand: DuPont™
Manufacturer Part Number: D13398700
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11713023
Quantity Price
1 £ 585.0 / Pack of 100
Estimated Shipment
12-06-2017
Add to basket
Description and Specification
Specification
Size X-Large
Size (Metric) Chest 108 to 116cm
Protection Class Category III, Type 5 and 6, EN 1073-2
Waist Style Elastic
Product Type Hooded Coverall
Style Hooded
Material Microporous Film
Color White
Ankle Style Elastic
For Use With (Application) For that are less demanding in terms of barrier, durability and comfort, for example general maintenance, hospitals and other industries. Spraying or any work involving composite materials, glass manufacturing, and utilities
Seam Type Stitched (internal)