DuPont™ Microporous film Hooded Coverall
Overview
Brand: DuPont™
Manufacturer Part Number: D13398835
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11763123
Quantity Price
1 £ 585.0 / Pack of 100
Estimated Shipment
15-05-2017
Add to basket
Description and Specification
Specification
Protection Class Category III, Type 5 and 6, EN 1073-2
Size Medium
Size (Metric) Chest 92 to 100cm
Waist Style Elastic
Style Hooded
Product Type Hooded Coverall
Ankle Style Elastic
Material Microporous Film
Color White
For Use With (Application) For that are less demanding in terms of barrier, durability and comfort, for example general maintenance, hospitals and other industries. Spraying or any work involving composite materials, glass manufacturing, and utilities
Seam Type Stitched (internal)