missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ MicroPlus Membranes
Features very narrow pore size distribution, low levels of extractables and increased temperature stability. Whatman™ MicroPlus Membranes are suitable for sample preparation, microbiological studies and filtration of aqueous solutions.
Brand: Cytiva 10407170
Code : 53
Additional Details : Weight : 0.63000kg
Description
The membrane filtration (MF) method is excellent for the determination and enumeration of microbial contaminants.- Special membrane filters made from cellulose nitrate developed for microbiological quality control
- High mechanical strength
- High flow rate
- Sterile, individually packed
- All membrane filters have a high-contrast grid (3.1 mm)
- Type STL in dispenser boxes for easy removal and secure handling with the Membrane-Butler
Specifications
Green | |
Cellulose Nitrate | |
0.45 μm | |
Microbiological quality control | |
31mm | |
47 mm |
Membrane Butler | |
MicroPlus-41STL | |
Sterile | |
Black | |
Grid |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Flammable solid Category 1