missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ MicroPlus Membranes

Features very narrow pore size distribution, low levels of extractables and increased temperature stability. Whatman™ MicroPlus Membranes are suitable for sample preparation, microbiological studies and filtration of aqueous solutions.

Brand:  Cytiva 10407714

Code : 53

Additional Details : Weight : 0.11000kg

 View more versions of this product

Product Code. 11919217

  • £158.00 / Pack of 100
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

The membrane filtration (MF) method is excellent for the determination and enumeration of microbial contaminants.
  • Special membrane filters made from cellulose nitrate developed for microbiological quality control
  • High mechanical strength
  • High flow rate
  • Sterile, individually packed
  • All membrane filters have a high-contrast grid (3.1 mm)
  • Type STL in dispenser boxes for easy removal and secure handling with the Membrane-Butler
Specifications

Specifications

White
MicroPlus-21ST
Sterile
Black
Grid
Cellulose Nitrate
0.45 μm
Microbiological quality control
31mm
50 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers