Learn More
Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Integrate these microplate carriers with the microplate swinging bucket rotor.
£837.24
Specifications
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10188734 View Documents Thermo Scientific™
75003625
Sealed Microplate Carriers £837.24
Each View alternative products
Save Up to 
»
Estimated Shipment
25-06-2020
10034024 View Documents Thermo Scientific™
75003625
Sealed Microplate Carriers £837.24
2 sets View alternative products
Save Up to 
»
In Stock 17
Description
Description
Includes:
Sealed carriers include trays and lids (set of 2). Replacement O-rings for lids are offered as a set of four, with grease.
Specifications
Specifications
Biocontainment certified by CAMR™, HPA, Porton Down, U.K.