missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Yellow Plastic Buckets and Wringers

Brand:  Micronova™ W-2

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13168645

  • £129.63 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

0.47 to 0.94L
Yellow
Buckets
Wringer
16 to 32 oz., 0.47 to 0.94L
Cleanroom support applications and general janitorial use
Plastic
Each
Videos