missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Scoops

Brand:  Micronova™ SCP-01

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13137656

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Scoop
1100 mL
Autoclavable
37.19 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers