Learn More
Micronova™ PowerHead™ Disposable Mop Heads
Available in several different types and densities of foam and material coverings
Brand:  Micronova™ PH6-154
Code : 04
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11912485
  • £18.49 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Suitable for use on floors and walls. Complete mop consists of a PowerHead disposable mop head, a PowerHead handle (sold separately) and an extendible or single-length stainless-steel handle (sold separately).
PowerHead mop types include:
Yellow foam
  • Heavy-duty
  • Two distinct cleaning surfaces
Dense Urethane Foam
  • Disposable sponge mop head for cleanroom support areas
  • Two distinct cleaning surfaces
  • Withstands strong disinfectants
Ether and Dense Ether Foam
  • Ideal for applying detergents and disinfectants
  • Dense absorbent foam for aseptic and sterile support areas
  • Two distinct cleaning surfaces
Wide-Pore Foam
  • Economical, absorbent foam for pharmaceutical and sterile support areas
  • Suited for detergent and disinfectant applications
  • Two distinct cleaning surfaces
NovaPoly™ Covered Foam
  • Polyester covered to minimize particulations
  • For critical applications, pharmaceutical and sterile support facilities
Order Info
All mop heads are autoclavable and lauderable. Handle is autoclavable.
Specifications
Specifications
Sterile
NovaPoly covered foam
Yes
Yes
PowerHeads™ NovaPoly™ Mop Head
Use in pharmaceutical and sterile support facilities
Videos