Micronova™ PowerHead™ Bar
Overview
Brand: Micronova™
Manufacturer Part Number: PHB-14
UNSPSC: 47131600
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 13177486
Quantity Price
1 £ 240.0 / Each
Estimated Shipment
18-05-2017
Add to basket
Description and Specification
Specification
Material Stainless Steel
For Use With (Equipment) QD-PHH powerhead handles
For Use With (Application) For cleaning in cleanrooms, pharmaceutical environments, research and animal labs
Product Type PowerHeads™ Bar
Autoclavable Yes