Learn More
Micronova™ PCX™ Polyethylene Tapes
Low-density polyethylene tapes low in halogens and leachable chlorides
Brand:  Micronova™ PCX-34WH
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13108515
  • £18.50 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Perfect choice for color coding, sealing, labeling, masking, and other cleanroom tasks requiring pressure-sensitive qualities
  • Cleanroom processed and packaged
  • Polyethylene film and acrylic adhesive lessen chance of outgassing and offer excellent chemical resistance and good conformability
Order Info
Available in rolls of 33m (36 yd.) in several colors and widths
Contact your Customer Service Representative at 1-800-772-6733 for ordering information on additional colors and widths.
Specifications
Specifications
Non-sterile
FDA
33m
LDPE (Low-density Polyethylene)
Each
White
Sealing, color coding, labeling, batching and food processing applications
Tape
Cleanroom Tape
Videos