Learn More
Micronova™ MicroLoop™ Mop Heads
Brand: Micronova™ ML-9
Code : 03
Additional Details : Weight : 1.00000kg
Description
• ISO Class 5 to 8 (Class 100 to 100,000)
• Durable looped construction
Sturdy and functional looped mop heads intended for use in service areas outside of cleanrooms.
ML-1 MicroLoop PVA mop head is ideal for clean-build projects, post-construction, and cleanroom support. ML-9 MicroLoop Polyester mop head is an economical choice for hazardous materials cleanup and general spill control.
Order Info
Attach to a stainless-steel handle (sold separately) with 19-035-362B Thread Adapter.
Compliance
Both mop heads are rated for ISO Class 5 to 8 (Class 100 to 100,000) environments.
Specifications
MicroLoop Mop | |
Autoclavable | |
Ideal for clean build projects, post construction, cleanroom support | |
Polyester |