missing translation for 'onlineSavingsMsg'
Learn More

Swab on stick 44mm x 19mm x 44mm

Brand:  Micronova FS-175

Swab on stick 44mm x 19mm x 44mm

Code : 03

Additional Details : Weight : 1.00000kg

Product Code. 13167786

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.