Learn More
Micronova™ CR100 PC™ Cleanroom Tape—White
Processed for applications in and around cleanrooms
Brand:  Micronova™ CR100PC-12RE
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12813632
  • £11.17 / Each
Estimated Shipment
18-06-2018
Add to basket
Description
Description
  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag
Specifications
Specifications
Red
Matte
33m
VinylĂľRubber, synthetic
Each
FDA, CID
Sealing, color coding, labeling, batching and food processing applications
Tape
Cleanroom Tape
No
Videos