missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ CR100 PC™ Cleanroom Tape—Dark Blue

Processed for applications in and around cleanrooms

Brand:  Micronova™ CR100PC-12DB

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12853622

  • £11.17 / Each


Estimated Shipment
22-06-2018
Add to basket

Description

Description

  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag

Specifications

Specifications

Dark Blue
Matte
33m
Vinyl
Each
PPP-T-66E, CID A-A 1689A
Sealing, color coding, labeling, batching
Tape
Cleanroom Tape
No
Videos