Learn More
Millipore™ Microfilm Funnel and Filter
Brand:  Millipore™ MIAAWG100
Code : 72
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13178053
  • £216.00 / Pack of 150
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Convenient, easy to use system for well accepted membrane filtration method conforms to international standards: CEN, Standard Methods, WHO
  • Disposable, stackable, light, recyclable plastic Microfil™ funnels eliminate autoclaving and minimise waste
  • 'Push fit' Microfil™ funnels tightly seal to manifold supports without clamps or O-rings assuring leak-free operation and uniform micro-organism recoveries
  • Novel device lifts membrane after filtration, for easy removal with forceps
  • Unique Microfil™ support filters faster than all-glass filter apparatus
  • 100mL or 250mL funnels available with convenient accessory dispensers
  • Choose conventional sterile membranes or novel EZ-Pak™ membrane cartridges. Select manifold-mounted filtration supports for laboratory use or stand-alone on-site filtration support.
Specifications
Specifications
White Gridded
100mL
100mL
150Pack
150Pack
Polypropylene/Mixed Cellulose Esters (MCE)
Microfilm Funnel and Filter, 100mL, 47mm dia. Filter, White Gridded, Polypropylene/Mixed Cellulose Esters (MCE) 0.8μm Pore Size, MF-Millipore Media, Sterile, for Yeast or mold Applications, Clinical Lab, Environmental Analysis, Industrial QC, Food and Beverage
Videos
Documentation
Technical Documents