Learn More
Emma Safety Footwear Micro Safety Shoes
Unique microfiber upper is fully washable. SmartecAir™ provides fresh dry feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors used in high-tech and cleanroom environments.
Brand: Emma Safety Footwear 500540-44
Additional Details : Weight : 1.44700kg
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
White | |
Hydro-Tec™ Sanitized Silver | |
44 |
Elastic | |
Micro | |
Microfiber | |
Steel |