Learn More
Technical Service Consultants™ Micro-organism Storage
Facilitate the integrity of your bacterial preservation system using Technical Service Consultants™ Micro-organism Storage. This system enables you to reduce the labor costs of continual sub-culturing and eliminates the need to purchase expensive freeze drying equipment. The culture viability can be checked regularly without sacrificing the entire vial.
Brand:  Technical Service Consultants™ TS/71-GN
Code : 03
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 11574129
  • £204.00 / Pack of 100
Estimated Shipment
15-06-2018
Add to basket
Description
Description
  • 100-vial capacity - available in range of colors
  • Contains 20 to 25 treated beads in cryopreservation fluid
  • Long-term storage of::bacteria, fungi and yeasts; quality control cultures; culture collections; and cross-infection isolates
Specifications
Specifications
Green
Bacterial bead preservation system in polypropylene moisture-resistant freezer box/tray
Videos
Documentation
Technical Documents
View accessory products