Learn More
Eppendorf™ Micro Fusion Chamber
Easy to use. Integrated cuvette chamber and extremely high transformation rates
Brand:  Eppendorf™ 4308030003
Code : 99
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 12816532
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Extremely high transformation rates due to innovative electronics and buffers
  • Integrated cuvette chamber
  • Easy to use
  • Compact design
  • Universal electroporator for eukaryotic, yeast and bacteria
Specifications
Specifications
For Fusion
1 Ea.
0.2 mm
Micro Fusion Chamber
1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products