missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II, APC, clone: YKIX334.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5909-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15578156

  • £65.25 / 25 tests

Estimated Shipment
30-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This YKIX334.2 monoclonal antibody reacts with canine major histocompatibility complex (MHC) Class II. The MHC plays a significant role in immunity by presenting self and non-self antigens to T cells. In dogs, MHC Class II is expressed on all T and B cells, macrophages, fibroblasts, and kidney interstitial cells.This monoclonal antibody has been reported to inhibit mixed lymphocyte reactions and T cell activation induced by MHC Class II antigen presentation in vitro. Also, YKIX334.2 recognition of MHC Class II is not allele-specific.Applications Reported: This YKIX334.2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This YKIX334.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal canine peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class II
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Major histocompatibility complex class II, MHC 2, MHC Class II, MHC II, MHCCII
IgG2a, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Canine
Flow Cytometry
YKIX334.2
APC
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
Documents

For Research Use Only.