missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II I-Ab, PE, clone: AF6-120.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5320-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537006

  • £49.60 / 25µg


Estimated Shipment date
from Supplier 29-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This AF6-120.1.2 monoclonal antibody reacts with the mouse MHC Class II I-Ab alloantigen of H-2b bearing mouse strains, including C57Bl/6 and 129. This cell surface molecule is involved in antigen presentation to T cells expressing CD3/TCR and CD4.The AF6-120.1 antibody crossreacts with H-2k and H-2u haplotypes. However, reactivity to other haplotypes (i.e., d, f, g7, p, q, r, and s) has not been observed.Applications Reported: This AF6-120.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This AF6-120.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class II I-Ab
AF6-120.1
PE
Conjugated
IgG2a
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos