Learn More
MHC Class II I-Ad Monoclonal Antibody (AMS-32.1), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 17-5323-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This AMS-32.1 monoclonal antibody reacts with the mouse MHC Class II I-Ad alloantigen, which is present on mouse strains such as Balb/c and DBA/2. This antibody also recognizes the H-2f, H-2g7, H-2i, and H-2v haplotypes. However, reactivity to other haplotypes (e.g., b, k, p, q, r, s, and u) has not been observed. Applications Reported: This AMS-32.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AMS-32.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
MHC Class II I-Ad Monoclonal antibody specifically detects MHC Class II I-Ad in Mouse samples. It is validated for Flow Cytometry.Specifications
MHC Class II I-Ad | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01921, P06342, P06343, P06344, P06345, P06346, P14483 | |
H2-Ab1 | |
Affinity chromatography | |
RUO | |
14961 | |
4° C, store in dark, DO NOT FREEZE! | |
IgG2b κ |
Flow Cytometry | |
AMS-32.1 | |
APC | |
H2-Ab1 | |
Abeta; AI845868; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; Ia2; Ia-2; IAb; I-Abeta; major histocompatibility complex class II beta chain; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Liquid |
For Research Use Only.