missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II I-Ad, APC, clone: AMS-32.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5323-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15330740

  • £49.53 / 25µg


Estimated Shipment
27-07-2018
Add to basket

Description

Description

Description: This AMS-32.1 monoclonal antibody reacts with the mouse MHC Class II I-Ad alloantigen, which is present on mouse strains such as Balb/c and DBA/2. This antibody also recognizes the H-2f, H-2g7, H-2i, and H-2v haplotypes. However, reactivity to other haplotypes (e.g., b, k, p, q, r, s, and u) has not been observed.Applications Reported: This AMS-32.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This AMS-32.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class II (I-A)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06342
H-2 class II histocompatibility antigen A beta chain, H2-AB1, H2-iabeta
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
AMS-32.1
APC
H2-Ab1
Conjugated
H2-Ab1
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14961
Videos

For Research Use Only.