missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II (I-A/I-E), eVolve 655, clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 86-5321-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15570837

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Description: The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.Applications Reported: This M5/114.15.2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This M5/114.15.2 antibody has been pre-titrated and tested by flow cytometric analysis of mouse splenocytes. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 655 can be optimally excited with laser lines from 325-405 nm, and emits near 655 nm. We recommend using the violet (405 nm) laser line for excitation and a 630LP with a 660/40 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-635 nm; Emission: 657 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

MHC Class II (I-A/I-E)
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P04231, P06342
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
M5/114.15.2
eVolve 655
H2-Ab1
Conjugated
H2-Ab1, H2-Eb1
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary
14961, 14969
Videos
Documents

For Research Use Only.