missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II (I-A/I-E), clone: M5/114.15.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-5321-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227417

  • £66.65 / 100µg


EU Stock 2
Add to basket

Description

Description

Description: The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.Applications Reported: This M5/114.15.2 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), immunohistology staining of frozen tissue sections, immunohistology staining of paraffin embedded tissue sections and in vitro blocking of T cell proliferative responses. (Please use Functional Grade purified M5/114.15.2, cat.16-4321, in functional assays.).Applications Tested: The M5/114.15.2 antibody has been tested by flow cytometric analysis of mouse splenocytes and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class II (I-A/I-E)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06342, P04231
I-Ab, I-Ad, I-Aq, I-Ed, I-Ek, MHC class 2, MHC class II glygoproteins
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryþFunctional AssayþImmunohistochemistryþImmunohistochemistry (Frozen)þImmunoprecipitationþWestern Blot
M5/114.15.2
Unconjugated
H2-Ab1
Purified
H2-Ab1, H2-Eb1
IgG2b, kappa
100μg
Store at 2-8°C.
Primary
14961, 14969
Videos

For Research Use Only.