missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (RT1-A) Mouse anti-Rat, FITC, Clone: OX18, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0921-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15576456

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This OX18 monoclonal antibody recognizes a non-polymorphic determinant of rat MHC Class I, which is also known as RT1-A. Rat MHC Class I is expressed on all nucleated cells and erythrocytes.

The OX18 antibody has been reported to have functional activity.
Specifications

Specifications

MHC Class I (RT1-A)
OX18
FITC
Conjugated
IgG1, kappa
RUO
Primary
Rat
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.