missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (RT1-A) Mouse anti-Rat, FITC, Clone: OX18, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0921-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15586456

  • £125.00 / 100µg

Estimated Shipment
02-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This protein is a non-polymorphic determit of rat MHC Class I, which is also known as RT1-A. Rat MHC Class I is expressed on all nucleated cells and erythrocytes.
Specifications

Specifications

MHC Class I (RT1-A)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Liquid
RT1-A
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100188935
Flow Cytometry
OX18
FITC
RT1-A
rat class I A
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Videos
Documents

For Research Use Only.