missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (H-2Kk), eFluor 450, clone: AF3-12.1.3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5940-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15331810

  • £56.31 / 25µg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This AF3-12.1.3 monoclonal antibody reacts with the H-2Kk MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, f, j, k, p, q, r, s, u, and v) has not been observed.Applications Reported: This AF3-12.1.3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This AF3-12.1.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class I (H-2Kk)
AF3-12.1.3
eFluor 450
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MHC I, H2Kk, H2-Kk, H2K-k
IgG1
RUO
Primary
Murine
Videos