Learn More
MHC Class I (H-2Kb) Monoclonal Antibody (AF6-88.5.5.3), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£113.00 - £276.00
Specifications
Antigen | MHC Class I (H-2Kb) |
---|---|
Clone | AF6-88.5.5.3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15311380
|
Affymetrix eBioscience
46-5958-82 |
100 μg |
£276.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15301380
|
Affymetrix eBioscience
46-5958-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This AF6-88.5.5.3 monoclonal antibody reacts with the H-2Kb MHC class I alloantigen. H-2Kb is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. d, f, j, k, p, q, r, s, u, and v) has not been observed. Applications Reported: This AF6-88.5.5.3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF6-88.5.5.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative.
MHC Class I (H-2Kb) Monoclonal antibody specifically detects MHC Class I (H-2Kb) in Mouse samples. It is validated for Flow Cytometry.Specifications
MHC Class I (H-2Kb) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
14972 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
AF6-88.5.5.3 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Mouse | |
H2-K1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
H2-K1 |
For Research Use Only.