missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (H-2Kd), eFluor 450, clone: SF1-1.1.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5957-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371810

  • £62.06 / 25µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: This SF1-1.1.1 monoclonal antibody reacts with the H-2Kd MHC class I alloantigen. H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, j, k, p, q, s, and v) has not been observed.Applications Reported: This SF1-1.1.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This SF1-1.1.1 antibody has been tested by flow cytometric analysis of mouse splecnocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class I (H-2Kd)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01902
H-2K, H-2K(d), H2-K, MGC184092, MGC7052
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
SF1-1.1.1
eFluor 450
H2-K1
Conjugated
H2-K1
IgG2a
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14972
Videos

For Research Use Only.