missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (H-2Kd/H-2Dd), eFluor 450, clone: 34-1-2S, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5998-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15559786

  • £62.63 / 25µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p.Applications Reported: This 34-1-2S antibody has been reported for use in flow cytometric analysis.Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class I (H-2Kd/H-2Dd)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01902, P14426
H-2 class I histocompatibility antigen, K-D alpha chain, H2K1, H2KD, H2DD
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
34-1-2S
eFluor 450
H2-D1
Conjugated
H2-D1, H2-K1
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14964, 14972
Videos

For Research Use Only.