missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (H-2Kd/H-2Dd), Biotin, clone: 34-1-2S, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5998-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239999

  • £49.99 / 50µg


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p.Applications Reported: 34-1-2S has been reported for use in flow cytometric analysis.Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MHC Class I (H-2Kd/H-2Dd)
34-1-2S
Biotin
Conjugated
IgG2a
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos