Learn More
MHC Class I (H-2Kd/H-2Dd) Monoclonal Antibody (34-1-2S), Biotin, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£108.00 - £229.00
Specifications
Antigen | MHC Class I (H-2Kd/H-2Dd) |
---|---|
Clone | 34-1-2S |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15249999
|
Affymetrix eBioscience
13-5998-82 |
100 μg |
£108.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15259999
|
Affymetrix eBioscience
13-5998-85 |
500 μg |
£229.00
500µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 34-1-2S monoclonal antibody reacts with the mouse MHC class I, H-2Kd and H-2Dd. This cytotoxic antibody also cross-reacts with Kb, s, r, q, p. Applications Reported: 34-1-2S has been reported for use in flow cytometric analysis. Applications Tested: This 34-1-2S antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.Specifications
MHC Class I (H-2Kd/H-2Dd) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
14964, 14972 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
34-1-2S | |
Flow Cytometry | |
Biotin | |
Mouse | |
Mouse | |
H2-D1, H2-K1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.