Learn More
mGluR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 720375
Additional Details : Weight : 0.01000kg
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Pig, Rabbit and Cat.
mGluR4 Polyclonal antibody specifically detects mGluR4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
mGluR4 | |
Polyclonal | |
Unconjugated | |
GRM4 | |
G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; GPRC1D; GRM4; Metabotropic glutamate receptor 4; mGlu4; MGLUR4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
Q14833 | |
GRM4 | |
Peptide corresponding to Human GRM4 (aa 33-46 ) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |