missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MGEA5 (Human) Recombinant Protein
Human MGEA5 full-length ORF ( NP_036347.1, 1 a.a. - 916 a.a.) recombinant protein with GST-tag at N-terminal.
£280.00 - £425.00
Specifications
Accession Number | NP_036347.1 |
---|---|
Gene ID (Entrez) | 10724 |
Name | meningioma expressed antigen 5 (hyaluronidase) |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16145602
|
Abnova™
H00010724-P01.10UG |
10 μg |
£280.00
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16155602
|
Abnova™
H00010724-P01.25UG |
25 μg |
£425.00
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MVQKESQATLEERESELSSNPAASAGASLEPPAAPAPGEDNPAGAGGAAVAGAAGGARRFLCGVVEGFYGRPWVMEQRKELFRRLQKWELNTYLYAPKDDYKHRMFWREMYSVEEAEQLMTLISAAREYEIEFIYAISPGLDITFSNPKEVSTLKRKLDQVSQFGCRSFALLFDDIDHNMCAADKEVFSSFAHAQVSITNEIYQYLGEPETFLFCPTEYCGTFCYPNVSQSPYLRTVGEKLLPGIEVLWTGPKVVSKEIPVESIEEVSKIIKRAPVIWDNIHANDYDQKRLFLGPYKGRSTELIPRLKGVLTNPNCEFEANYVAIHTLATWYKSNMNGVRKDVVMTDSEDSTVSIQIKLENEGSDEDIETDVLYSPQMALKLALTEWLQEFGVPHQYSSRQVAHSGAKASVVDGTPLVAAPSLNATTVVTTVYQEPIMSQGAALSGEPTTLTKEEEKKQPDEEPMDMVVEKQEETDHKNDNQILSEIVEAKMAEELKPMDTDKESIAESKSPEMSMQEDCISDIAPMQTDEQTNKEQFVPGPNEKPLYTAEPVTLEDLQLLADLFYLPYEHGPKGAQMLREFQWLRANSSVVSVNCKGKDSEKIEEWRSRAAKFEEMCGLVMGMFTRLSNCANRTILYDMYSYVWDIKSIMSMVKSFVQWLGCRSHSSAQFLIGDQEPWAFRGGLAGEFQRLLPIDGANDLFFQPPPLTPTSKVYTIRPYFPKDEASVYKICREMYDDGVGLPFQSQPDLIGDKLVGGLLSLSLDYCFVLEDEDGICGYALGTVDVTPFIKKCKISWIPFMQEKYTKPNGDKELSEAEKIMLSFHEEQEVLPETFLANFPSLIKMDIHKKVTDPSVAKSMMACLLSSLKANGSRGAFCEVRPDDKRILEFYSKLGCFEIAKMEGFPKDVVILGRSL
Specifications
NP_036347.1 | |
meningioma expressed antigen 5 (hyaluronidase) | |
125% SDS-PAGE Stained with Coomassie Blue | |
FLJ11229, FLJ23355, KIAA0679, MEA5, NCOAT, OGA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
10724 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGEA5 | |
GST |