Learn More
MG 624, Tocris Bioscience™
α7 neuronal nicotinic receptor antagonist
Brand:  Tocris (Bio-Techne) 135610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13469213
  • £148.47 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
A selective antagonist for neuronal α7 subunit- containing nAChR subtypes. Inhibits α-Bgtx binding to chick α7 and α4β2 subtypes with Ki values of 106 nM and 84 μM respectively.
Specifications
Chemical Identifiers
77257-42-2
451.392
stilonium iodide, stilonio ioduro dcit, stilonii iodidum inn-latin, iodure de stilonium inn-french, ioduro de estilonio inn-spanish, 2-p-styrylphenoxy ethyl triethylammonium iodide, triethyl 2-p-styrylphenoxy ethyl ammonium iodide, ioduro di trietil-beta-4-stilbenossi-etil ammonio italian, ammonium, 2-p-styrylphenoxy ethyl triethyl-, iodide, dsstox_cid_25781
triethyl-[2-[4-[(E)-2-phenylethenyl]phenoxy]ethyl]azanium;iodide
C22H30INO
RDTKUZXIHMTSJO-UEIGIMKUSA-M
6433339
CC[N+](CC)(CC)CCOC1=CC=C(C=C1)C=CC2=CC=CC=C2.[I-]
Specifications
10mg
77257-42-2
stilonium iodide, stilonio ioduro dcit, stilonii iodidum inn-latin, iodure de stilonium inn-french, ioduro de estilonio inn-spanish, 2-p-styrylphenoxy ethyl triethylammonium iodide, triethyl 2-p-styrylphenoxy ethyl ammonium iodide, ioduro di trietil-beta-4-stilbenossi-etil ammonio italian, ammonium, 2-p-styrylphenoxy ethyl triethyl-, iodide, dsstox_cid_25781
CC[N+](CC)(CC)CCOC1=CC=C(C=C1)C=CC2=CC=CC=C2.[I-]
451.392
451.39
MG 624
C22H30INO
RDTKUZXIHMTSJO-UEIGIMKUSA-M
triethyl-[2-[4-[(E)-2-phenylethenyl]phenoxy]ethyl]azanium;iodide
6433339
>98%