missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MG 132
Click to view available options
Quantity:
1mg
5mg
10mg
Description
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.
Solubility
Soluble in ethanol, chloroform, methanol, DMSO.
Notes
Protect from heat. Store away from oxidizing agents.
MG 132 is used to inhibitor of calpain proteases, a proteasome and NF-κB inhibitor used as a tool for perturbing the proteasome-regulated degradation of intracellular proteins.
Solubility
Soluble in ethanol, chloroform, methanol, DMSO.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Color | White |
Content And Storage | Store at -20°C |
Format | Crystalline Solid |
Molecular Weight (g/mol) | 475.63 |
Product Type | MG 132 |
Quantity | 1mg |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'