missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566112
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: DQRAERILHKE QGKTGEVIKL TDVKDFS Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.
MFSD1 Polyclonal antibody specifically detects MFSD1 in Human samples. It is validated for Immunocytochemistry.Specifications
MFSD1 | |
Polyclonal | |
Unconjugated | |
MFSD1 | |
1200003O06Rik; hypothetical protein LOC641486; major facilitator superfamily domain containing 1; major facilitator superfamily domain-containing protein 1; Major facilitator superfamily domain-containing protein 1-like protein; Mfsd1; mfsd1a; RGD1305751; SMAP4; SMAP-4; Smooth muscle cell-associated protein 4; UG0581B09; zgc:123187 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3U5 | |
MFSD1 | |
Recombinant Human MFSD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only