missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFRP Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | MFRP |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse MFRP Gln91-Pro584 Accession # Q8K480 |
Gene ID (Entrez) | 83552 |
Gene Accession No. | Q8K480 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
MFRP Polyclonal specifically detects MFRP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MFRP | |
83552 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) MFRP is observed, 15% cross-reactivity with recombinant mouse (rm) Frizzled-4 is observed, 10% cross-reactivity with rmFrizzled-1, rhFrizzled-5, rmFrizzled-8, and rmFrizzled-10 is observed, and 5% cross-reactivity with rmFrizzled-3, rmFrizzled-6, and rmFrizzled-7 is observed. | |
Unconjugated | |
Western Blot, Immunohistochemistry | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant mouse MFRP Gln91-Pro584 Accession # Q8K480 | |
Q8K480 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only