missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFRP C-Terminus Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | MFRP |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
MFRP Polyclonal specifically detects MFRP in Human samples. It is validated for Western Blot.Specifications
MFRP | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6 | |
MFRP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9BY79 | |
83552 | |
Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579 Accession # Q9BY79 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human MFRP C-Terminus in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-4, recombinant human Frizzled-5, rmFrizzled-6, rmFrizzled-7, rmFrizzled-8, and rmFrizzled-9 is observed. |
For Research Use Only