missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFNG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561842
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TTRAFHRLRL ELLLDTWVSR TREQTFVFTD SPDKGLQERL GSHLVVTNCS AEHSHPALSC KMAAEFDTFL AS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.
MFNG Polyclonal antibody specifically detects MFNG in Human samples. It is validated for Immunohistochemistry.Specifications
MFNG | |
Polyclonal | |
Unconjugated | |
MFNG | |
AW546563; beta-1,3-N-acetylglucosaminyltransferase manic fringe; EC 2.4.1.222; manic fringe homolog; Mfng; MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4242 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00587 | |
MFNG | |
Recombinant protein corresponding to Human MFNG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only