missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ F1 Premium Cylindrical Knob Weights

Used in mass labs all over the world, and for calibration of analytical and micro balances

Brand:  Mettler Toledo™ 00158640

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12713165

  • £92.36 / Each


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Initial weight calibration uses robotic technology which excludes human error and leads to consistent and reproducible results with low uncertainty values Premium, vacuum melted steel offers an anticorrosive surface, and low magnetization and susceptibility values Monobloc design and electro polishing of surface provide maximum stability
Specifications

Specifications

Weight with knob, One piece (monobloc)
OIML Class F1
20g
20g
Wooden box
1
ISO17025, ISO9001, ISO14001 environmental standard
Stainless Steel
8kg/dm3 at 20°C
Cylindrical
Each
Videos