Learn More
Mettler Toledo™ Solutions for perfectION™ Combined ISE
Reference electrolytes for ion selective electrodes.
Brand:  Mettler Toledo™ 51344750
Code : 59
Additional Details : CAS Number : 7761-88-8 Weight : 0.30000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
 View more versions of this product
Product Code. 12631296
  • £93.20 / 300mL
Estimated Shipment
12-06-2018
Add to basket
Description
Description
Reference filling solution.
  • For perfectION Combined ion selective electrodes
  • 5 x 60mL (each)
Reference Electrolytes
  • Ion Electrolyte A (calcium, fluoride)
  • Ion Electrolyte B (chloride, cyanide, lead, silver/sulfide)
  • Ion Electrolyte C (silver)
  • Ion Electrolyte D (copper, iodide)
  • Ion Electrolyte E (potassium)
  • Ion Electrolyte F (nitrate)
Specifications
Specifications
Ion Electrolyte A (calcium, fluoride)
Combined ISE Solution for perfectION
Videos