Learn More
Mettler Toledo™ SG8 SevenGo pro™ pH/Ion Meter
Built-in data memory and automatic functions for outstanding performance
Brand:  Mettler Toledo™ 51302582
Code : 59
Additional Details : Weight : 0.34000kg
 View more versions of this product
Product Code. 10700275
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Portable meter measures ion concentration, pH, mV and temperature
  • Designed for easy, comfortable right- or left-handed operation
  • Choice of three packaging configurations
  • Waterproof and dustproof to IP-67 standard
  • Large backlit display of measured value and settings
  • Rotatable electrode clip that mounts left or right for single-handed operation
  • IP-67 pH/Ion meter with expanded range for the highest demands
  • Continuous indication of electrode and battery status
  • Automatic endpoint and buffer recognition
  • Automatic temperature compensation
  • Three-point calibration with predefined or user-defined buffers
  • Segmented or linear calibration as desired
  • Five-point ion calibration with choice of 6 predefined and one user-defined buffer set
  • Record date, time, time interval
  • Intuitive menu operation using softkeys
  • GLP memory for 200 datapoints
  • Contact-free infrared communication with notebook, printer or PC
  • ON/OFF backlit display
Specifications
Specifications
pH/Ion Meter, InLab 413 SG electrode (1.8m cable), four AA 1.5V batteries/1.3V NiMH accumulator
0.1/0.01/0.001
0 to 1999mV
0 to 9999ppm
5
±0.2°C
200
IR to printer or PC via RS232 or USB
325g
21.9cm
8.89 cm
-2 to 20
±0.01
0.1mV
3/4 Digit
0.1°C
LCD
BNC (IP67), Cinch (IP67), NTC 30kilohm
1.75 x 3.5 x 8.62 in. (4.5 x 8.89 x 21.9cm)
Measures ion concentration, pH, mV, temp
4.5cm
Electrode Meter Kit with pH electrode
Videos
Documentation View accessory products