missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenGo™ perfectION Combined ISE Membrane Modules

Brand:  Mettler Toledo™ 51344852

UNSPSC : 41115608

Code : 04

Additional Details : Weight : 0.88000kg

 View more versions of this product

Product Code. 11907941

  • £231.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Can flow easily through the junction and junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled
ISA
PerfectION Combined NO-3 ISE
Lemo
0° to 40°C
Combined ISE Membrane module