Mettler Toledo™ SevenGo™ perfectION Combined ISE Electrodes

Get an “All-In-One Solution” with SevenGo™ perfectION Combined ISE Electrodes. The brilliant Click & Clear™ press mechanism allows for quick cleaning and ensures stable readings even in difficult samples.

Overview
Brand: Mettler Toledo™

Manufacturer Part Number: 51344818

UNSPSC: 41115608

Code: 04

Additional Details:
Additional Details: Weight: 0.33000kg



Product Code. 11917831

Quantity Price
1 £ 835.0 / Each
Estimated Shipment
11-05-2017
Add to basket


Description and Specification

Specification

Filling Solution Type ISA
Electrode Combined ISE Electrode
Connector Types Lemo
Applications I- ions
Operating Range (Metric) 0.005 to 127000mg/L
Temperature (Metric) Operating 0° to 80°C

  • As vast as the range of different ions, is the selection of ion­selective electrodes (ISE) 
  • The special Click & Clear™ junction allows for an optimal contact of the electrolyte solution and the sample
  • With the dedicated solutions the sample can be optimally prepared for successful measurement of ion concentration