missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenExcellence™ S500 pH/Ion Meters

Intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. Mettler Toledo SevenExcellence S500 pH/Ion Meter is modularly expandable with additional measurement parameters.

Brand:  Mettler Toledo™ 30046248

Code : 03

Additional Details : Weight : 1.98000kg

 View more versions of this product

Product Code. 13460679

  • £1720.00 / Each


Estimated Shipment
12-03-2020
Add to basket

Description

Description

  • Measures pH, conductivity and dissolved oxygen
  • Methods for incremental ion measurements, guiding the user through the individual steps
  • Large 7 in. color display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • Optional comprehensive qualification package including IQ/OQ/PQ
Specifications

Specifications

SevenExcellence pH/Ion meter S500
-2 to 20
±0.002
0.1/1mV
0 to 999999mg/L
0.001μS/cm to 2000mS/cm
0.001μS/cm to 2000mS/cm
0.001μS/cm to 2000mS/cm
0.001μS/cm to 2000mS/cm
Max. 5 points, 8 pre defined and 20 user defined buffer group
13 Pre defined and 20 user defined standard
0.1°C
Single-channel
20000
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
±0.5%
0.001 to 999999mg/L ppm, 0.0001 to 100mol/L %, 0.001 to 100000mmol/L
1 pH/ion module and 2 blank modules, electrode arm, calibration report and declaration of conformity, installation instructions and Quick guide also on CD-ROM, 2 x Sachet packs for pH 2, pH 7, pH 9.21, pH 10
0.001/0.01/0.1
-2000 to 2000mV
±0.1mV
0.001 to 1
0.001 to 1
0.001 to 1
0.001 to 1
0.001 to 1
Max. 5 points, 8 pre defined and 20 user defined buffer group
-30° to +130°C
±0.1°C
TFT
250 analyses (20.000 data points)
RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
S500-BASIC
Videos
Documents