SevenExcellence™ S400 pH/mV Meters

Intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. Mettler Toledo SevenExcellence S400 pH/mV Meter is a single-channel instrument which is modularly expandable with additional measurement parameters at any time — conductivity, ion or dissolved oxygen.

Overview
Brand: Mettler Toledo Online

Manufacturer Part Number: 30046243

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 1.98000kg



Product Code. 13400679

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Data Points 20000
Range (ISE) 0 to 0.999mg/L
Range (Conductivity) 0.001μS/cm to 2000mS/cm
Resolution (mV) 0.1/1mV
Calibration Points (ISE) Max. 5 points, 8 pre defined and 20 user defined buffer group
Range (mV) -2000 to 2000mV
Model S400-ISFET
Outputs USB, RS232 and Ethernet
Display Type TFT
Resolution (Temperature) 0.1°C
Accuracy (pH) ±0.002
Resolution (ISE) 0.001 to 1
Resolution (pH) 0.001/0.01/0.1
Resolution (Conductivity) 0.001 to 1
Calibration Points (Conductivity) 13 Pre defined and 20 user defined standard
Calibration Points (pH) Max. 5 points, 8 pre defined and 20 user defined buffer group
Accuracy (Temperature) ±0.1°C
Range (pH) -2 to 20
Includes S-400 Meter; InLab 490; guide to pH measurements; 2 buffer sachets for pH 4.01, 7, 9.21 and 10
Accuracy (mV) ±0.1mV
Range (Temperature) -30° to +130°C