missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenEasy and SevenMulti™ pH Meter Accessories

Accessories for use with SevenEasy and SevenMulti Series meters.

Brand:  Mettler Toledo™ 51302819

Code : 59

Additional Details : Weight : 0.12000kg

 View more versions of this product

Product Code. 10111532

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

For use with SevenEasy and SevenMulti Series meters.
Specifications

Specifications

SevenEasy™ and SevenMulti™ pH Meter
pH
Protective Cover
Videos